Polyethylene glycol monooleyl ether n=∼20
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
P0715-25G
P0715-500G
TCIAP0715-25GEA
132
DKK
TCIAP0715-25G
TCIAP0715-500G
Polyethylene glycol monooleyl ether n=∼20
Polyethylene glycol monooleyl ether
Formel:
HO(CH₂CH₂O)nC₁₈H₃₅ |
MDL Number:
MFCD00074369 CAS-nummer: 9004-98-2 EINECS: 500-016-2 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to lump |
Acid Value | max. 1.0 |
The average value of n | 16.0 to 23.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...