Poly(vinyl formal)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Polyvinylformal, Formvar

P0641-500G P0641-25G
TCIAP0641-500GEA 952 DKK
TCIAP0641-500G TCIAP0641-25G
Poly(vinyl formal)
Poly(vinyl formal)
Formel: (C₃H₆O₂)n
MDL Number: MFCD00084463
CAS-nummer: 63450-15-7

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange granule
Vinyl formal min. 80.0 %
Volatile matter Content max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR