Vinyl propionate ≥98.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
P0613-25ML
P0613-500ML
TCIAP0613-25MLEA
140
DKK
TCIAP0613-25ML
TCIAP0613-500ML
Vinyl propionate ≥98.0% (by GC) stabilised
Vinyl propionate
Formel:
C₅H₈O₂ Kogepunkt: 95°C Flammepunkt: 8°C |
MDL Number:
MFCD00027004 CAS-nummer: 105-38-4 EINECS: 203-293-5 UN nummer: 3272 ADR: 3,II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...