Pontacyl violet 6R

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P0593-1G P0593-5G
TCIAP0593-1GEA 698 DKK
TCIAP0593-1G TCIAP0593-5G
Pontacyl violet 6R
Pontacyl violet 6R
Formel: C₂₀H₁₄N₂O₈S₂
MDL Number: MFCD00059522
CAS-nummer: 5850-63-5

Bestil nu

Specifikationer Testresultater

Appearance Gray to Dark purple to Black powder to crystal
Lambda max. 538.0 to 542.0 nm (H2O)
Absorbance (E1%1cm) min. 470 (H2O, 538.0 ~ 542.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR