Pyrogallol red

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3',4',5',6'-Tetrahydroxyspiro[benzo[c][1,2]oxathiole-3,9'-xanthene] 1,1-dioxide

P0572-1G P0572-5G
TCIAP0572-1GEA 238 DKK
TCIAP0572-1G TCIAP0572-5G
Pyrogallol red
Pyrogallol red
Formel: C₁₉H₁₂O₈S
Molekylvægt: 400.36 g/mol
MDL Number: MFCD00005046
CAS-nummer: 32638-88-3
EINECS: 251-134-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e14f0a9d974372efca47aa4ca013fd9b5326e53&vid=f7cae98194e1103fd1adbf052246c1ddf0eaf3cd&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Dark red to Brown powder to crystal
Lambda max. 539.0 to 543.0 nm (pH 8.0 buffer sol.)
Absorbance (E1%1cm) min. 850 (pH 8.0 buffer sol., 539.0 to 543.0 nm)
Sensitiveness Colorimetric Test : Turn Purple with Nickel and turn Red by adding EDTA 2Na


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR