Tricresylphosphater (blanding af isomerer) ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Tricresyl phosphate, Tritolyl phosphate, Tris(methylphenyl) phosphate, TCP
P0273-25G
P0273-500G
TCIAP0273-25GEA
148
DKK
TCIAP0273-25G
TCIAP0273-500G
Tricresylphosphater (blanding af isomerer) ≥99.0% (by GC)
Tricresylphosphater (blanding af isomerer)
Formel:
C₂₁H₂₁O₄P Molekylvægt: 368.37 g/mol Kogepunkt: 420 Smeltepunkt: -40 Flammepunkt: 234°C |
MDL Number:
MFCD02102116 CAS-nummer: 1330-78-5 EINECS: 215-548-8 UN nummer: 3082 ADR: 9 III Merck Index: 14,09763 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0% (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...