3-Oxo-4-aza-5alpha-androstan-17beta-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

O0514-1G O0514-5G
TCIAO0514-1GEA 608 DKK
TCIAO0514-1G TCIAO0514-5G
3-Oxo-4-aza-5alpha-androstan-17beta-carboxylic acid ≥98.0% (by HPLC, titration analysis)
3-Oxo-4-aza-5alpha-androstan-17beta-carboxylic acid
Formel: C₁₉H₂₉NO₃
Smeltepunkt: 315°C
MDL Number: MFCD06411020
CAS-nummer: 103335-55-3
EINECS: 407-820-1

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +73.0 to +81.0 deg (C=1, pyridine)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR