Octadecyl rhodamine B chloride
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
6-(Diethylamino)-N,N-diethyl-9-[2-[(octadecyloxy)carbonyl]phenyl]-3H-xanthen-3-iminium chloride, 3,6-Bis(diethylamino)-9-[2-[(octadecyloxy)carbonyl]phenyl]xanthylium chloride, N-[6-(Diethylamino)-9-[2-[(octadecyloxy)carbonyl]phenyl]-3H-xanthen-3-ylidene]-N-ethylethanaminium chloride
O0512-10MG
O0512-50MG
TCIAO0512-10MGEA
1150
DKK
TCIAO0512-10MG
TCIAO0512-50MG
Octadecyl rhodamine B chloride
Octadecyl rhodamine B chloride
Formel:
C₄₆H₆₇ClN₂O₃ Molekylvægt: 731.50 g/mol |
MDL Number:
MFCD00467892 CAS-nummer: 65603-19-2 |
Specifikationer Testresultater
Appearance | Orange to Amber to Dark red powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | gt. 3.5 to 4.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...