3,4-Dihydro-3-oxo-2H[1,4]benzothiazin-2ylacetic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
O0504-1G
O0504-5G
TCIAO0504-1GEA
444
DKK
TCIAO0504-1G
TCIAO0504-5G
3,4-Dihydro-3-oxo-2H[1,4]benzothiazin-2ylacetic acid ≥98.0% (by HPLC, titration analysis)
3,4-Dihydro-3-oxo-2H[1,4]benzothiazin-2ylacetic acid
Formel:
C₁₀H₉NO₃S |
MDL Number:
MFCD00087524 CAS-nummer: 6270-74-2 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...