2-Oxo-2H-chromen-7-yltrifluormethanesulphonate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 7-Coumaryl triflate, Trifluoromethanesulphonic acid 2-Oxo-2H-chromen-7-yl ester, 2-Oxo-2H-chromen-7-yl trifluoromethanesulphonate, 7-Coumaryl trifluoromethanesulphonate

O0472-1G O0472-5G
TCIAO0472-1GEA 1190 DKK
TCIAO0472-1G TCIAO0472-5G
2-Oxo-2H-chromen-7-yltrifluormethanesulphonate ≥98.0% (by GC)
2-Oxo-2H-chromen-7-yltrifluormethanesulphonate
Formel: C₁₀H₅F₃O₅S
Molekylvægt: 294.21 g/mol
MDL Number: MFCD18382236
CAS-nummer: 108530-10-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af289dac9a13e58356df20f2b8776fdd9d1d3efb&vid=af289dac9a13e58356df20f2b8776fdd9d1d3efb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 81.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR