3-Oxetanon ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
O0467-1G
O0467-5G
TCIAO0467-1GEA
394
DKK
TCIAO0467-1G
TCIAO0467-5G
3-Oxetanon ≥97.0% (by GC)
3-Oxetanon
Formel:
C₃H₄O₂ Kogepunkt: 106°C Flammepunkt: 26°C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD09263255 CAS-nummer: 6704-31-0 UN nummer: 1993 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...