1-(4-Chlorobenzyl)-1H-indole-3-carbaldehyde ≥95.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-(4-Chlorobenzyl)-1H-indole-3-carboxaldehyde, Oncrasin 1, 1-[(4-Chlorophenyl)methyl]-1H-indole-3-carboxaldehyde, Oncrasin-1

O0450-10MG O0450-50MG
TCIAO0450-10MGEA 337 DKK
TCIAO0450-10MG TCIAO0450-50MG
1-(4-Chlorobenzyl)-1H-indole-3-carbaldehyde ≥95.0% (by HPLC)
1-(4-Chlorobenzyl)-1H-indole-3-carbaldehyde
Formel: C₁₆H₁₂ClNO
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01051808
CAS-nummer: 75629-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8eca242af88f69afd9b4bfe58b9245608e0abfcf&vid=8eca242af88f69afd9b4bfe58b9245608e0abfcf&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 4.90 to 5.50 %
Melting point 115.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR