Oxfendazole ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl (6-(phenylsulphinyl)-1H-benzo[d]imidazol-2-yl)carbamate, Methyl [5-(Phenylsulphinyl)benzimidazol-2-yl]carbamate, [5-(Phenylsulphinyl)benzimidazol-2-yl]carbamic acid Methyl Ester

O0391-1G O0391-5G
TCIAO0391-1GEA 238 DKK
TCIAO0391-1G TCIAO0391-5G
Oxfendazole ≥98.0% (by HPLC, total nitrogen)
Oxfendazole
Formel: C₁₅H₁₃N₃O₃S
MDL Number: MFCD00801063
CAS-nummer: 53716-50-0
EINECS: 258-714-5
Merck Index: 14,06935

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR