7-Octen-1-ol ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

O0280-25ML O0280-5ML
TCIAO0280-25MLEA 4260 DKK
TCIAO0280-25ML TCIAO0280-5ML
7-Octen-1-ol ≥96.0% (by GC)
7-Octen-1-ol
Formel: C₈H₁₆O
Kogepunkt: 66°C
MDL Number: MFCD00798076
CAS-nummer: 13175-44-5
EINECS: 236-122-8

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR