3-Octen-2-one ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
O0252-25ML
O0252-5ML
TCIAO0252-25MLEA
460
DKK
TCIAO0252-25ML
TCIAO0252-5ML
3-Octen-2-one ≥96.0% (by GC)
3-Octen-2-one
Formel:
C₈H₁₄O Molekylvægt: 126.20 g/mol |
MDL Number:
MFCD00015565 CAS-nummer: 1669-44-9 |
|
Specifikationer Testresultater
Appearance | Colorless to Yellow to Green clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...