Octanenitrile ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Heptyl Cyanide

O0221-25ML O0221-250ML
TCIAO0221-25MLEA 329 DKK
TCIAO0221-25ML TCIAO0221-250ML
Octanenitrile ≥97.0% (by GC)
Octanenitrile
Formel: C₈H₁₅N
Molekylvægt: 125.22 g/mol
Kogepunkt: 200°C
Flammepunkt: 73°C
MDL Number: MFCD00001983
CAS-nummer: 124-12-9
EINECS: 204-682-2

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR