Trimethyl orthobutyrate ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
O0179-25ML
O0179-5ML
TCIAO0179-25MLEA
1100
DKK
TCIAO0179-25ML
TCIAO0179-5ML
Trimethyl orthobutyrate ≥96.0% (by GC)
Trimethyl orthobutyrate
Formel:
C₇H₁₆O₃ Kogepunkt: 146°C Flammepunkt: 35°C |
MDL Number:
MFCD00008480 CAS-nummer: 43083-12-1 EINECS: 256-079-9 UN nummer: 3272 ADR: 3,III |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...