γ-Oryzanol

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

O0172-25G O0172-250G
TCIAO0172-25GEA 238 DKK
TCIAO0172-25G TCIAO0172-250G
γ-Oryzanol
gamma-Oryzanol
Formel: C₄₀H₅₈O₄
MDL Number: MFCD00867548
CAS-nummer: 11042-64-1
UN nummer: 2811
ADR: 6.1,II
Merck Index: 14,06886

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystaline
Lambda max. 313.0 to 317.0 nm (Heptane)
Absorbance min. 0.520 (15mg/L, Heptane, 313.0 to 317.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR