Oxamic acid potassium salt ≥98.0% (by total nitrogen and titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

O0169-25G O0169-500G
TCIAO0169-25GEA 542 DKK
TCIAO0169-25G TCIAO0169-500G
Oxamic acid potassium salt ≥98.0% (by total nitrogen and titration analysis)
Oxamic acid potassium salt
Formel: C₂H₃NO₃
MDL Number: MFCD00058475
CAS-nummer: 21141-31-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) 98.0 to 102.0 %
Purity (Nonaqueous Titration) 98.0 to 102.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR