Potassium oxonate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Potassium Allantoxanate, Oxonic acid Potassium Salt, Potassium 4,6-Dihydroxy-1,3,5-triazine-2-carboxylate, Allantoxanic acid Potassium Salt, Potassium 4,6-dioxo-1,4,5,6-tetrahydro-1,3,5-triazine-2-carboxylate

O0164-25G O0164-5G
TCIAO0164-25GEA 731 DKK
TCIAO0164-25G TCIAO0164-5G
Potassium oxonate ≥98.0% (by HPLC, titration analysis)
Potassium oxonate
Formel: C₄H₃N₃O₄
MDL Number: MFCD00010565
CAS-nummer: 2207-75-2
EINECS: 218-627-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR