2,3,5-Trimethyl-4-nitropyridine N-Oxide ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

N1189-1G N1189-250MG
TCIAN1189-1GEA 887 DKK
TCIAN1189-1G TCIAN1189-250MG
2,3,5-Trimethyl-4-nitropyridine N-Oxide ≥98.0% (by GC, titration analysis)
2,3,5-Trimethyl-4-nitropyridine N-Oxide
Formel: C₈H₁₀N₂O₃
MDL Number: MFCD00800254
CAS-nummer: 86604-79-7

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 76.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR