Neochlorogenic acid ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5-O-(trans-3,4-Dihydroxycinnamoyl)-D-quinic acid, trans-5-O-Caffeoylquinic acid

N1155-10MG N1155-50MG
TCIAN1155-10MGEA 911 DKK
TCIAN1155-10MG TCIAN1155-50MG
Neochlorogenic acid ≥98.0% (by HPLC)
Neochlorogenic acid
Formel: C₁₆H₁₈O₉
Opbevaringstemperatur: Køleskab
MDL Number: MFCD10566639
CAS-nummer: 906-33-2
EINECS: 212-997-1

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Specific rotation [a]20/D +4.0 to +8.0 deg (C=0.2, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR