2-(2-Nitrophenyl)propan-1-ol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

N1133-1G N1133-5G
TCIAN1133-1GEA 2090 DKK
TCIAN1133-1G TCIAN1133-5G
2-(2-Nitrophenyl)propan-1-ol ≥98.0% (by GC)
2-(2-Nitrophenyl)propan-1-ol
Formel: C₉H₁₁NO₃
MDL Number: MFCD09038679
CAS-nummer: 64987-77-5

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR