5-Nitrobenzofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
N1035-1G
N1035-5G
TCIAN1035-1GEA
509
DKK
TCIAN1035-1G
TCIAN1035-5G
5-Nitrobenzofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)
5-Nitrobenzofuran-2-carboxylic acid
Formel:
C₉H₅NO₅ Smeltepunkt: 275°C |
MDL Number:
MFCD00060513 CAS-nummer: 10242-12-3 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...