Nebivolol hydrochloride ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

N0954-20MG N0954-100MG
TCIAN0954-20MGEA 673 DKK
TCIAN0954-20MG TCIAN0954-100MG
Nebivolol hydrochloride ≥98.0% (by HPLC)
Nebivolol hydrochloride
Formel: C₂₂H₂₅F₂NO₄
MDL Number: MFCD07357273
CAS-nummer: 152520-56-4
Merck Index: 14,06432
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6de78a678b8b50348efe6955076b771ac93da38&vid=c6de78a678b8b50348efe6955076b771ac93da38&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 223.0 to 228.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR