7-Nitro-1-tetralon ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,4-Dihydro-7-nitronaphthalen-1(2H)-one, 7-Nitro-3,4-dihydronaphthalen-1(2H)-one

N0945-1G N0945-5G
TCIAN0945-1GEA 222 DKK
TCIAN0945-1G TCIAN0945-5G
7-Nitro-1-tetralon ≥98.0% (by GC)
7-Nitro-1-tetralon
Formel: C₁₀H₉NO₃
MDL Number: MFCD00019661
CAS-nummer: 40353-34-2
EINECS: 254-887-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b3707199f94bc56ebe6f088401c53eb5768446&vid=77b3707199f94bc56ebe6f088401c53eb5768446&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR