Nateglinide ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
N-[(trans-4-Isopropylcyclohexyl)carbonyl]-D-phenylalanine, (R)-2-rel-((1r,4R)-4-Isopropylcyclohexanecarboxamido)-3-phenylpropanoic acid
N0912-500MG
N0912-5G
N0912-1G
TCIAN0912-500MGEA
271
DKK
TCIAN0912-500MG
TCIAN0912-5G
TCIAN0912-1G
Nateglinide ≥98.0% (by HPLC, titration analysis)
Nateglinide
Formel:
C₁₉H₂₇NO₃ |
MDL Number:
MFCD00875706 CAS-nummer: 105816-04-4 Merck Index: 14,06428 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -36.5 to -40.0 deg (C=1, 0.1mol/L NaOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...