2-Nitroimidazol ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

N0891-1G N0891-5G
TCIAN0891-1GEA 1190 DKK
TCIAN0891-1G TCIAN0891-5G
2-Nitroimidazol ≥98.0% (by HPLC, titration analysis)
2-Nitroimidazole
Formel: C₃H₃N₃O₂
Smeltepunkt: 288°C
MDL Number: MFCD00005185
CAS-nummer: 527-73-1
EINECS: 208-425-5
UN nummer: 2811
ADR: 6.1,III
Merck Index: 14,00921
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7989383f8eec457d182d79d78b4f18359ca33e80&vid=7989383f8eec457d182d79d78b4f18359ca33e80&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR