4-Nitrobenzyl acetoacetate ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

N0875-25G N0875-5G
TCIAN0875-25GEA 1010 DKK
TCIAN0875-25G TCIAN0875-5G
4-Nitrobenzyl acetoacetate ≥98.0% (by HPLC)
4-Nitrobenzyl acetoacetate
Formel: C₁₁H₁₁NO₅
MDL Number: MFCD00799237
CAS-nummer: 61312-84-3

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 42.0 to 46.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR