Nobiletin ≥95.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3',4',5,6,7,8-Hexamethoxyflavone, 2-(3,4-Dimethoxyphenyl)-5,6,7,8-tetramethoxy-4H-chromen-4-one

N0871-10MG N0871-100MG
TCIAN0871-10MGEA 993 DKK
TCIAN0871-10MG TCIAN0871-100MG
Nobiletin ≥95.0% (by HPLC)
Nobiletin
Formel: C₂₁H₂₂O₈
Opbevaringstemperatur: Fryser
MDL Number: MFCD03273560
CAS-nummer: 478-01-3

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 136.0 to 140.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR