Nicorandil ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

N0837-1G N0837-5G N0837-100MG
TCIAN0837-1GEA 575 DKK
TCIAN0837-1G TCIAN0837-5G TCIAN0837-100MG
Nicorandil ≥98.0% (by HPLC, titration analysis)
Nicorandil
Formel: C₈H₉N₃O₄
MDL Number: MFCD00186520
CAS-nummer: 65141-46-0
EINECS: 265-514-1
Merck Index: 14,06521
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f2535b429068e6893b2e1481144e92e417a9ced&vid=0f2535b429068e6893b2e1481144e92e417a9ced&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 88.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR