2-Nitro-p-cymene ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

N0801-25G N0801-5G
TCIAN0801-25GEA 3780 DKK
TCIAN0801-25G TCIAN0801-5G
2-Nitro-p-cymene ≥95.0% (by GC)
2-Nitro-p-cymene
Formel: C₁₀H₁₃NO₂
Kogepunkt: 134°C
MDL Number: MFCD00007176
CAS-nummer: 943-15-7
EINECS: 213-397-2

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR