Nonaethylene glycol monomethyl ether ≥93.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2,5,8,11,14,17,20,23,26-Nonaoxaoctacosan-28-ol, mPEG9-Alcohol

N0699-1G N0699-500MG
TCIAN0699-1GEA 821 DKK
TCIAN0699-1G TCIAN0699-500MG
Nonaethylene glycol monomethyl ether ≥93.0% (by GC)
Nonaethylene glycol monomethyl ether
Formel: C₁₉H₄₀O₁₀
Molekylvægt: 428.52 g/mol
Smeltepunkt: 14°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06797148
CAS-nummer: 6048-68-6

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR