(R)-Ethylpiperidine-3-carboxylate-(2R,3R)-2,3-dihydroxysuccinate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (R)-Ethyl piperidine-3-carboxylate (2S,3S)-2,3-dihydroxysuccinate, Ethyl (R)-Nipecotate L-Tartrate, (R)-Nipecotic acid ethyl ester L-Tartrate, (R)-3-Piperidinecarboxylic acid ethyl ester L-Tartrate, Ethyl (R)-nipecotate L-tartrate, Ethyl (R)-3-Piperidinecarboxylate L-Tartrate

N0680-25G N0680-5G
TCIAN0680-25GEA 1710 DKK
TCIAN0680-25G TCIAN0680-5G
(R)-Ethylpiperidine-3-carboxylate-(2R,3R)-2,3-dihydroxysuccinate ≥98.0% (by titrimetric analysis)
(R)-Ethylpiperidine-3-carboxylate-(2R,3R)-2,3-dihydroxysuccinate
Formel: C₁₂H₂₁NO₈
Molekylvægt: 307.30 g/mol
MDL Number: MFCD00799538
CAS-nummer: 167392-57-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 153.0 to 158.0 deg-C
Specific rotation [a]20/D +9.5 to +11.0 deg (C=5, H2O)
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR