Nicotinamide-N-oxide ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Nicotinamide N-oxide, Pyridine-3-carboxamide N-oxide, 3-Carbamoylpyridine 1-oxide
N0669-25G
N0669-5G
TCIAN0669-25GEA
2620
DKK
TCIAN0669-25G
TCIAN0669-5G
Nicotinamide-N-oxide ≥98.0% (by titrimetric analysis)
Nicotinamide-N-oxide
Formel:
C₆H₆N₂O₂ Molekylvægt: 138.13 g/mol |
MDL Number:
MFCD00006202 CAS-nummer: 1986-81-8 EINECS: 217-859-4 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in hot Water | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...