Nonadecanedioic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
N0663-1G
N0663-5G
TCIAN0663-1GEA
1090
DKK
TCIAN0663-1G
TCIAN0663-5G
Nonadecanedioic acid ≥98.0% (by titrimetric analysis)
Nonadecanedioic acid
Formel:
C₁₉H₃₆O₄ |
MDL Number:
MFCD00039536 CAS-nummer: 6250-70-0 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light red powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 119.0 to 123.0 deg-C |
Solubility in EtOH | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...