1,2-Nonanediol ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

N0651-5G N0651-1G
TCIAN0651-5GEA 1880 DKK
TCIAN0651-5G TCIAN0651-1G
1,2-Nonanediol ≥99.0% (by GC)
1,2-Nonanediol
Formel: C₉H₂₀O₂
Kogepunkt: 155°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01861291
CAS-nummer: 42789-13-9

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 35.0 to 38.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR