Sodium-2-naphthoate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

N0617-25G N0617-5G
TCIAN0617-25GEA 1360 DKK
TCIAN0617-25G TCIAN0617-5G
Sodium-2-naphthoate ≥98.0% (by titrimetric analysis)
Sodium-2-naphthoate
Formel: C₁₁H₈O₂
MDL Number: MFCD00671570
CAS-nummer: 17273-79-9

Bestil nu

Specifikationer Testresultater

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR