Nonafluorovaleric acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

N0605-25G N0605-5G
TCIAN0605-25GEA 2040 DKK
TCIAN0605-25G TCIAN0605-5G
Nonafluorovaleric acid ≥98.0% (by GC, titration analysis)
Nonafluorovaleric acid
Formel: C₅HF₉O₂
Kogepunkt: 70°C
MDL Number: MFCD00040211
CAS-nummer: 2706-90-3
EINECS: 220-300-7
UN nummer: 3265
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR