1,2-Naphthalenedicarbonitrile ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1,2-Dicyanonaphthalene

N0578-1G N0578-5G
TCIAN0578-1GEA 911 DKK
TCIAN0578-1G TCIAN0578-5G
1,2-Naphthalenedicarbonitrile ≥98.0% (by GC)
1,2-Naphthalenedicarbonitrile
Formel: C₁₂H₆N₂
Molekylvægt: 178.19 g/mol
MDL Number: MFCD00191673
CAS-nummer: 19291-76-0
UN nummer: 3439
ADR: 6.1 III

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 191.0 to 195.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR