3,3',3''-Nitrilotris(propionamide) ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

N0342-25G N0342-500G
TCIAN0342-25GEA 296 DKK
TCIAN0342-25G TCIAN0342-500G
3,3',3''-Nitrilotris(propionamide) ≥98.0% (by titrimetric analysis)
3,3',3''-Nitrilotris(propionamide)
Formel: C₉H₁₈N₄O₃
Molekylvægt: 230.27 g/mol
MDL Number: MFCD00059194
CAS-nummer: 2664-61-1
EINECS: 220-196-3

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 182.0 to 186.0 deg-C
Solubility in hot Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR