4-Nitrotoluene-2-sulphonic acid hydrate ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: p-Nitrotoluene-o-sulphonic acid

N0277-25G N0277-500G
TCIAN0277-25GEA 279 DKK
TCIAN0277-25G TCIAN0277-500G
4-Nitrotoluene-2-sulphonic acid hydrate ≥95.0% (by titrimetric analysis)
4-Nitrotoluene-2-sulphonic acid hydrate
Formel: C₇H₇NO₅S
Molekylvægt: 217.20 g/mol
MDL Number: MFCD00024914
CAS-nummer: 121-03-9
EINECS: 204-445-3
UN nummer: 3261
ADR: 8 III

Bestil nu

Specifikationer Testresultater

Appearance White to Brown powder to crystal
Purity (Neutralization titration) min. 95.0 % (calcd.on anh.substance) (calcd. on exclusive of SO4)
Water max. 20.0 %
Sulfate max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR