5-Nitrobenzimidazole nitrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

N0151-25G N0151-250G
TCIAN0151-25GEA 197 DKK
TCIAN0151-25G TCIAN0151-250G
5-Nitrobenzimidazole nitrate ≥98.0% (by titrimetric analysis)
5-Nitrobenzimidazole nitrate
Formel: C₇H₅N₃O₂
MDL Number: MFCD00013157
CAS-nummer: 27896-84-0
EINECS: 248-716-4

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR