Sodium-1-naphthol-8-sulphonate ≥96.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Sodium 8-Hydroxy-1-naphthalenesulphonate, Sodium 8-hydroxynaphthalene-1-sulphonate, 8-Hydroxy-1-naphthalenesulphonic acid Sodium Salt, 1-Naphthol-8-sulphonic acid Sodium Salt
N0034-25G
N0034-500G
TCIAN0034-25GEA
279
DKK
TCIAN0034-25G
TCIAN0034-500G
Sodium-1-naphthol-8-sulphonate ≥96.0% (by HPLC)
Sodium-1-naphthol-8-sulphonate
Formel:
C₁₀H₈O₄S Molekylvægt: 246.21 g/mol |
MDL Number:
MFCD00021524 CAS-nummer: 20215-36-5 EINECS: 243-605-7 |
Specifikationer Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...