Sodium-1-naphthaleneacetate ≥96.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
N0007-25G
N0007-500G
TCIAN0007-25GEA
304
DKK
TCIAN0007-25G
TCIAN0007-500G
Sodium-1-naphthaleneacetate ≥96.0% (by HPLC, titration analysis)
Sodium-1-naphthaleneacetate
Formel:
C₁₂H₁₀O₂ |
MDL Number:
MFCD00064967 CAS-nummer: 61-31-4 EINECS: 200-504-2 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Ion exchange titration) | min. 96.0 % (calcd.on dried substance) |
Solubility in Water | almost transparency |
Drying loss | max. 2.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...