3-Methyl-4-nitroquinoline 1-Oxide ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3-Methyl-4-nitroquinoline N-Oxide

M3311-1G M3311-250MG
TCIAM3311-1GEA 1500 DKK
TCIAM3311-1G TCIAM3311-250MG
3-Methyl-4-nitroquinoline 1-Oxide ≥98.0% (by HPLC, titration analysis)
3-Methyl-4-nitroquinoline 1-Oxide
Formel: C₁₀H₈N₂O₃
Molekylvægt: 204.19 g/mol
MDL Number: MFCD01685462
CAS-nummer: 14073-00-8

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 180.0 to 184.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR