Morpholine hydrobromide ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Morpholinium bromide

M3285-25G M3285-5G
TCIAM3285-25GEA 1280 DKK
TCIAM3285-25G TCIAM3285-5G
Morpholine hydrobromide ≥98.0% (by titrimetric analysis)
Morpholine hydrobromide
Formel: C₄H₉NO·HBr
Molekylvægt: 168.03 g/mol
MDL Number: MFCD22005717
CAS-nummer: 6377-82-8

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 210.0 to 214.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR