Monolinuron (N-(4-Chlorphenyl)-N'-methoxy-N'-methylcarbamid) ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M3269-25G M3269-5G
TCIAM3269-25GEA 1370 DKK
TCIAM3269-25G TCIAM3269-5G
Monolinuron (N-(4-Chlorphenyl)-N'-methoxy-N'-methylcarbamid) ≥98.0% (by HPLC, total nitrogen)
Monolinuron (N-(4-Chlorphenyl)-N'-methoxy-N'-methylcarbamid)
Formel: C₉H₁₁ClN₂O₂
MDL Number: MFCD00055264
CAS-nummer: 1746-81-2
EINECS: 217-129-5
UN nummer: 3077
ADR: 9,III

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR