2-(4-Methoxyphenyl)ethylamine hydrobromide ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Methoxyphenethylamine hydrobromide, 2-(4-Methoxyphenyl)ethylammonium bromide, 4-(2-Aminoethyl)anisole hydrobromide, 4-Methoxyphenethylammonium bromide
M3239-1G
M3239-5G
TCIAM3239-1GEA
206
DKK
TCIAM3239-1G
TCIAM3239-5G
2-(4-Methoxyphenyl)ethylamine hydrobromide ≥98.0% (by HPLC)
2-(4-Methoxyphenyl)ethylamine hydrobromide
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...