4-Methyl-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Methylpyridine-3-boronic acid pinacol ester, 2-(4-Methyl-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Methyl-3-pyridylboronic acid Pinacol ester, 4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

M3231-1G M3231-5G
TCIAM3231-1GEA 542 DKK
TCIAM3231-1G TCIAM3231-5G
4-Methyl-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-Methyl-3-pyridineboronic acid pinacol ester
Formel: C₁₂H₁₈BNO₂
Molekylvægt: 219.0878 g/mol
MDL Number: MFCD12923382
CAS-nummer: 1171891-31-8

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 106.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR